Search
Search
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013998435.2 | XP_013853889.1 | 287 | 94 | ||
| XM_013998436.2 | XP_013853890.1 | 409 | 94 | |||
| XM_013998437.2 | XP_013853891.1 | 441 | 94 | |||
| XM_013998438.2 | XP_013853892.1 | 484 | 94 | |||
| XM_013998439.2 | XP_013853893.1 | 1511 | 94 | |||
| XM_013998440.2 | XP_013853894.1 | 258 | 94 | |||
| XM_021097233.1 | XP_020952892.1 | 402 | 94 | |||
| XM_021097235.1 | XP_020952894.1 | 362 | 94 | |||
Set Membership: |
|