Search
Search
| Gene Symbol: CRACD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101353.1 | XP_020957012.1 | 633 | 105 | ||
| XM_021101354.1 | XP_020957013.1 | 735 | 105 | |||
| XM_021101355.1 | XP_020957014.1 | 549 | 105 | |||
| XM_021101356.1 | XP_020957015.1 | 507 | 105 | |||
| XM_021101358.1 | XP_020957017.1 | 548 | 105 | |||
Set Membership: |
|