Search
Search
| Gene Symbol: RHOT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131742.6 | XP_003131790.5 | 1225 | 126 | ||
| XM_003483060.3 | XP_003483108.2 | 1225 | 126 | |||
| XM_005656986.3 | XP_005657043.2 | 1225 | 126 | |||
| XM_013989958.2 | XP_013845412.2 | 1225 | 126 | |||
| XM_013989959.2 | XP_013845413.2 | 1225 | 126 | |||
| XM_013989961.2 | XP_013845415.2 | 1225 | 126 | |||
Set Membership: |
|