Search
Search
| Gene Symbol: ZC3HC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078727.1 | XP_020934386.1 | 683 | 67 | ||
| XM_021078728.1 | XP_020934387.1 | 532 | 67 | |||
| GenBank mRNA | AK233404.1 | - | 5 - 6 | 660 | 67 | |
| AK234184.1 | - | 5 - 6 | 486 | 67 | ||
Set Membership: |
|