Search
Search
| Gene Symbol: INTS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021101712.1 | XP_020957371.1 | 799 | 73 | ||
| GenBank mRNA | AK233521.1 | - | 5 - 6 | 774 | 73 | |
| AK343758.1 | - | 5 - 6 | 686 | 73 | ||
Set Membership: |
|