Search
Search
| Gene Symbol: ZBTB46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021077466.1 | XP_020933125.1 | 1624 | 65 | ||
| XM_021077467.1 | XP_020933126.1 | 1455 | 65 | |||
| XM_021077468.1 | XP_020933127.1 | 1783 | 65 | |||
Set Membership: |
|