Search
Search
| Gene Symbol: KIF13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100216.1 | XP_020955875.1 | 1432 | 125 | ||
| XM_021100217.1 | XP_020955876.1 | 1429 | 125 | |||
| XM_021100218.1 | XP_020955877.1 | 1432 | 125 | |||
| XM_021100219.1 | XP_020955878.1 | 1431 | 125 | |||
| XM_021100220.1 | XP_020955879.1 | 1432 | 125 | |||
| XM_021100222.1 | XP_020955881.1 | 1431 | 125 | |||
| XM_021100223.1 | XP_020955882.1 | 1431 | 125 | |||
| XM_021100224.1 | XP_020955883.1 | 1431 | 125 | |||
| XM_021100225.1 | XP_020955884.1 | 1431 | 125 | |||
| XM_021100226.1 | XP_020955885.1 | 1431 | 125 | |||
| XM_021100227.1 | XP_020955886.1 | 1431 | 125 | |||
| XM_021100228.1 | XP_020955887.1 | 1431 | 125 | |||
| XM_021100229.1 | XP_020955888.1 | 1431 | 125 | |||
| XM_021100230.1 | XP_020955889.1 | 1430 | 125 | |||
| XM_021100231.1 | XP_020955890.1 | 1430 | 125 | |||
Set Membership: |
|