Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06870954_m1
See other SYTL2 GE Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 19617287 - 19763439 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06870954_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_005667202.3
XP_005667259.2
3195
72
XM_005667203.3
XP_005667260.1
2698
72
XM_013979211.2
XP_013834665.2
7053
72
XM_013979213.2
XP_013834667.2
6971
72
XM_013979215.2
XP_013834669.2
7032
72
XM_013979216.2
XP_013834670.2
7005
72
XM_013979218.2
XP_013834672.2
5586
72
XM_013979222.2
XP_013834676.2
3216
72
XM_013979223.2
XP_013834677.2
3147
72
XM_013979224.2
XP_013834678.2
3126
72
XM_013979225.2
XP_013834679.2
3075
72
XM_013979226.2
XP_013834680.1
975
72
XM_013979227.2
XP_013834681.1
854
72
XM_021062588.1
XP_020918247.1
7323
72
XM_021062589.1
XP_020918248.1
6954
72
XM_021062590.1
XP_020918249.1
9175
72
XM_021062591.1
XP_020918250.1
6984
72
XM_021062592.1
XP_020918251.1
6933
72
XM_021062593.1
XP_020918252.1
6885
72
XM_021062594.1
XP_020918253.1
6765
72
XM_021062596.1
XP_020918255.1
5538
72
XM_021062597.1
XP_020918256.1
5473
72
XM_021062598.1
XP_020918257.1
5397
72
XM_021062599.1
XP_020918258.1
5229
72
XM_021062600.1
XP_020918259.1
5320
72
XM_021062601.1
XP_020918260.1
1118
72
XM_021062602.1
XP_020918261.1
1013
72
XM_021062603.1
XP_020918262.1
1006
72
XM_021062604.1
XP_020918263.1
2932
72
XM_021062605.1
XP_020918264.1
891
72
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Sus scrofa
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline