Search
Search
| Gene Symbol: CFAP45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089586.1 | XP_020945245.1 | 70 | 80 | ||
| XM_021089587.1 | XP_020945246.1 | 67 | 80 | |||
| XM_021089588.1 | XP_020945247.1 | 67 | 80 | |||
Set Membership: |
|