Search
Search

RAPGEF5| Gene Symbol: RAPGEF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656685.2 | XP_005656742.1 | 2240 | 104 | ||
| XM_005656686.2 | XP_005656743.1 | 2237 | 104 | |||
| XM_013989575.2 | XP_013845029.1 | 2365 | 104 | |||
| XM_013989576.2 | XP_013845030.1 | 2362 | 104 | |||
| XM_013989577.1 | XP_013845031.1 | 2363 | 104 | |||
| XM_021063440.1 | XP_020919099.1 | 2359 | 104 | |||
| XM_021063441.1 | XP_020919100.1 | 2192 | 104 | |||
| XM_021063442.1 | XP_020919101.1 | 2164 | 104 | |||
Set Membership: |
|