Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06874882_m1
See other FGGY GE Assays ›
Gene Symbol:
FGGY
Gene Name:
FGGY carbohydrate kinase domain containing
Gene Aliases:
-
Chromosome Location:
Chr.6: 152754923 - 153213179 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06874882_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: FGGY
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_021097787.1
XP_020953446.1
1371
78
XM_021097788.1
XP_020953447.1
1458
78
XM_021097789.1
XP_020953448.1
1696
78
XM_021097790.1
XP_020953449.1
1609
78
XM_021097792.1
XP_020953451.1
1466
78
XM_021097793.1
XP_020953452.1
1650
78
XM_021097794.1
XP_020953453.1
1370
78
XM_021097795.1
XP_020953454.1
1457
78
XM_021097796.1
XP_020953455.1
1695
78
XM_021097797.1
XP_020953456.1
1649
78
XM_021097798.1
XP_020953457.1
1370
78
XM_021097799.1
XP_020953458.1
1370
78
XM_021097800.1
XP_020953459.1
1371
78
XM_021097801.1
XP_020953460.1
1371
78
XM_021097803.1
XP_020953462.1
1370
78
XM_021097804.1
XP_020953463.1
1370
78
XM_021097805.1
XP_020953464.1
1370
78
XM_021097806.1
XP_020953465.1
1370
78
XM_021097807.1
XP_020953466.1
1370
78
XM_021097808.1
XP_020953467.1
1369
78
XM_021097809.1
XP_020953468.1
1369
78
XM_021097810.1
XP_020953469.1
1259
78
XM_021097811.1
XP_020953470.1
1259
78
GenBank mRNA
AK233605.1
-
9 - 10
1072
78
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
carbohydrate kinase
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit