Search
Search
| Gene Symbol: RCAN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013988974.2 | XP_013844428.1 | 1854 | 110 | ||
| XM_021098801.1 | XP_020954460.1 | 624 | 110 | |||
| XM_021098802.1 | XP_020954461.1 | 796 | 110 | |||
| GenBank mRNA | AK233736.1 | - | 3 - 4 | 612 | 110 | |
| AK350683.1 | - | 4 - 5 | 802 | 110 | ||
| AK390262.1 | - | 3 - 4 | 639 | 110 | ||
| AY609588.1 | - | 3 - 4 | 621 | 110 | ||
Set Membership: |
|