Search
Search
| Gene Symbol: IL10RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656637.3 | XP_005656694.1 | 281 | 100 | ||
| GenBank mRNA | AK233769.1 | - | 2 - 3 | 278 | 100 | |
| AK240564.1 | - | 2 - 3 | 244 | 100 | ||
Set Membership: |
|