Search
Search
| Gene Symbol: GABPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005663481.3 | XP_005663538.1 | 317 | 81 | ||
| GenBank mRNA | AK233787.1 | - | 1 - 2 | 176 | 81 | |
| AK235745.1 | - | 1 - 2 | 175 | 81 | ||
Set Membership: |
|