Search
Search
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927845.5 | XP_001927880.3 | 1710 | 62 | ||
| XM_021098594.1 | XP_020954253.1 | 1672 | 62 | |||
| XM_021098595.1 | XP_020954254.1 | 1670 | 62 | |||
| XM_021098596.1 | XP_020954255.1 | 1670 | 62 | |||
| XM_021098598.1 | XP_020954257.1 | 2028 | 62 | |||
| XM_021098599.1 | XP_020954258.1 | 3485 | 62 | |||
| GenBank mRNA | AK233842.1 | - | 7 - 8 | 1662 | 62 | |
Set Membership: |
|