Search
Search
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927845.5 | XP_001927880.3 | 2086 | 105 | ||
| XM_021098594.1 | XP_020954253.1 | 2048 | 105 | |||
| XM_021098595.1 | XP_020954254.1 | 2046 | 105 | |||
| XM_021098596.1 | XP_020954255.1 | 2046 | 105 | |||
| XM_021098598.1 | XP_020954257.1 | 2404 | 105 | |||
| XM_021098599.1 | XP_020954258.1 | 3861 | 105 | |||
| GenBank mRNA | AK233842.1 | - | 9 - 10 | 2038 | 105 | |
Set Membership: |
|