Search
Search
| Gene Symbol: KDM4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021063858.1 | XP_020919517.1 | 428 | 67 | ||
| XM_021063866.1 | XP_020919525.1 | 428 | 67 | |||
| GenBank mRNA | AK233909.1 | - | 2 - 3 | 328 | 67 | |
| AK345747.1 | - | 2 - 3 | 243 | 67 | ||
| AK348745.1 | - | 1 - 2 | 382 | 67 | ||
Set Membership: |
|