Search
Search
| Gene Symbol: DYM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092897.1 | XP_020948556.1 | 542 | 71 | ||
| XM_021092904.1 | XP_020948563.1 | 576 | 71 | |||
| XM_021092915.1 | XP_020948574.1 | 535 | 71 | |||
| XM_021092922.1 | XP_020948581.1 | 548 | 71 | |||
| XM_021092933.1 | XP_020948592.1 | 535 | 71 | |||
| XM_021092944.1 | XP_020948603.1 | 526 | 71 | |||
| XM_021092955.1 | XP_020948614.1 | 535 | 71 | |||
| GenBank mRNA | AK234991.1 | - | 3 - 4 | 488 | 71 | |
| AK400668.1 | - | 3 - 4 | 408 | 71 | ||
Set Membership: |
|