Search
Search
| Gene Symbol: MTF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481511.4 | XP_003481559.1 | 577 | 66 | ||
| XM_005663669.3 | XP_005663726.1 | 478 | 66 | |||
| XM_021090203.1 | XP_020945862.1 | 586 | 66 | |||
| XM_021090204.1 | XP_020945863.1 | 571 | 66 | |||
| XM_021090205.1 | XP_020945864.1 | 344 | 66 | |||
| XM_021090206.1 | XP_020945865.1 | 515 | 66 | |||
| XM_021090207.1 | XP_020945866.1 | 431 | 66 | |||
| GenBank mRNA | AK235445.1 | - | 4 - 5 | 465 | 66 | |
| AK239054.1 | - | 4 - 5 | 607 | 66 | ||
| AK239746.1 | - | 3 - 4 | 573 | 66 | ||
| AK347865.1 | - | 3 - 4 | 461 | 66 | ||
| AK351437.1 | - | 3 - 4 | 514 | 66 | ||
| AK395915.1 | - | 3 - 4 | 574 | 66 | ||
Set Membership: |
|