Search
Search
| Gene Symbol: PRMT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094135.1 | XP_020949794.1 | 1209 | 67 | ||
| XM_021094136.1 | XP_020949795.1 | 1236 | 67 | |||
| XM_021094137.1 | XP_020949796.1 | 1283 | 67 | |||
| XM_021094138.1 | XP_020949797.1 | 1267 | 67 | |||
| XM_021094139.1 | XP_020949798.1 | 1272 | 67 | |||
| XM_021094140.1 | XP_020949799.1 | 1305 | 67 | |||
| XM_021094141.1 | XP_020949800.1 | 1290 | 67 | |||
| XM_021094143.1 | XP_020949802.1 | 1287 | 67 | |||
| XM_021094144.1 | XP_020949803.1 | 1694 | 67 | |||
| XM_021094145.1 | XP_020949804.1 | 1272 | 67 | |||
| XM_021094146.1 | XP_020949805.1 | 1356 | 67 | |||
| XM_021094147.1 | XP_020949806.1 | 1291 | 67 | |||
| GenBank mRNA | AK235564.1 | - | 5 - 6 | 708 | 67 | |
| AK395119.1 | - | 3 - 4 | 470 | 67 | ||
| AK396944.1 | - | 9 - 10 | 1245 | 67 | ||
| AK398744.1 | - | 10 - 11 | 1240 | 67 | ||
Set Membership: |
|