Search
Search
| Gene Symbol: YBX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126475.3 | XP_003126523.1 | 1285 | 67 | ||
| XM_003355575.3 | XP_003355623.1 | 1080 | 67 | |||
| GenBank mRNA | AK235708.1 | - | 6 - 7 | 543 | 67 | |
| AK347123.1 | - | 1016 | 67 | |||
| AK396735.1 | - | 8 - 9 | 669 | 67 | ||
| AY610501.1 | - | 4 - 5 | 442 | 67 | ||
Set Membership: |
|