Search
Search
| Gene Symbol: ADGRE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083689.1 | XP_020939348.1 | 1656 | 98 | ||
| GenBank mRNA | AK230572.1 | - | 12 - 13 | 1620 | 98 | |
| AK230812.1 | - | 12 - 13 | 1521 | 98 | ||
Set Membership: |
|