Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Ss06880094_m1
See other ADGRE1 GE Assays ›
Gene Symbol:
ADGRE1
Gene Name:
adhesion G protein-coupled receptor E1
Gene Aliases:
EMR1
Chromosome Location:
Chr.2: 72217036 - 72305925 on Build Sscrofa11.1
Species:
Pig
Species Specific ID (Flybase ID):
-
Assay ID
Ss06880094_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
4,430.00
Your Price
Online offer:
4430.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ADGRE1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_021083951.1
XP_020939610.1
5604
79
XM_021083952.1
XP_020939611.1
5526
79
XM_021083953.1
XP_020939612.1
5457
79
XM_021083954.1
XP_020939613.1
5457
79
XM_021083955.1
XP_020939614.1
5457
79
XM_021083956.1
XP_020939615.1
5457
79
XM_021083957.1
XP_020939616.1
5457
79
XM_021083958.1
XP_020939617.1
5457
79
XM_021083959.1
XP_020939618.1
5457
79
XM_021083960.1
XP_020939619.1
5454
79
XM_021083962.1
XP_020939621.1
5448
79
XM_021083963.1
XP_020939622.1
5448
79
XM_021083964.1
XP_020939623.1
5448
79
XM_021083965.1
XP_020939624.1
5448
79
XM_021083966.1
XP_020939625.1
5448
79
XM_021083967.1
XP_020939626.1
5448
79
XM_021083968.1
XP_020939627.1
5448
79
XM_021083969.1
XP_020939628.1
5448
79
XM_021083970.1
XP_020939629.1
5409
79
XM_021083971.1
XP_020939630.1
5331
79
XM_021083972.1
XP_020939631.1
5328
79
XM_021083973.1
XP_020939632.1
2565
79
XM_021083974.1
XP_020939633.1
5379
79
GenBank mRNA
AK230628.1
-
18 - 19
2514
79
AK389644.1
-
14 - 15
1981
79
AK391528.1
-
15 - 16
2103
79
Back To Top
More Information
Set Membership:
Amplicon length between 71 and 85
Sus scrofa
Probe spans exons
Panther Classification:
Molecular Function -
G-protein coupled receptor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline