Search
Search
| Gene Symbol: BCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092208.1 | XP_020947867.1 | 1332 | 152 | ||
| XM_021092209.1 | XP_020947868.1 | 555 | 152 | |||
| XM_021092210.1 | XP_020947869.1 | 1019 | 152 | |||
| GenBank mRNA | AK230641.1 | - | 2 - 3 | 142 | 152 | |
Set Membership: |
|