Search
Search
| Gene Symbol: TRIP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021093813.1 | XP_020949472.1 | 883 | 105 | ||
| XM_021093818.1 | XP_020949477.1 | 883 | 105 | |||
| XM_021093826.1 | XP_020949485.1 | 670 | 105 | |||
| XM_021093833.1 | XP_020949492.1 | 995 | 105 | |||
| GenBank mRNA | AK230892.1 | - | 6 - 7 | 855 | 105 | |
| AK343920.1 | - | 6 - 7 | 855 | 105 | ||
Set Membership: |
|