Search
Search
| Gene Symbol: GON4L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089698.1 | XP_020945357.1 | 6261 | 97 | ||
| XM_021089699.1 | XP_020945358.1 | 6228 | 97 | |||
| XM_021089700.1 | XP_020945359.1 | 6013 | 97 | |||
| XM_021089701.1 | XP_020945360.1 | 6126 | 97 | |||
| XM_021089703.1 | XP_020945362.1 | 6216 | 97 | |||
| XM_021089704.1 | XP_020945363.1 | 6072 | 97 | |||
| XM_021089705.1 | XP_020945364.1 | 5937 | 97 | |||
| XM_021089706.1 | XP_020945365.1 | 6027 | 97 | |||
| XM_021089707.1 | XP_020945366.1 | 6219 | 97 | |||
| XM_021089708.1 | XP_020945367.1 | 4689 | 97 | |||
| GenBank mRNA | AK235950.1 | - | 6 - 7 | 859 | 100 | |
Set Membership: |
|