Search
Search
| Gene Symbol: FAM13A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100748.1 | XP_020956407.1 | 628 | 58 | ||
| GenBank mRNA | AK235993.1 | - | 3 - 4 | 600 | 58 | |
| AK346089.1 | - | 3 - 4 | 573 | 58 | ||
Set Membership: |
|