Search
Search

TMEM87A| Gene Symbol: TMEM87A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929220.5 | XP_001929255.2 | 1489 | 60 | ||
| XM_005659710.3 | XP_005659767.1 | 1514 | 60 | |||
| GenBank mRNA | AK236005.1 | - | 15 - 16 | 1439 | 60 | |
| AK391434.1 | - | 1 - 2 | 99 | 60 | ||
| AK393401.1 | - | 15 - 16 | 1514 | 60 | ||
| AK396673.1 | - | 15 - 16 | 1419 | 60 | ||
| AK399481.1 | - | 15 - 16 | 1466 | 60 | ||
Set Membership: |
|