Search
Search
| Gene Symbol: KARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126856.3 | XP_003126904.2 | 1646 | 91 | ||
| XM_003355743.3 | XP_003355791.1 | 1797 | 91 | |||
| GenBank mRNA | AK236350.1 | - | 12 - 13 | 1577 | 91 | |
| AK238649.1 | - | 13 - 14 | 1750 | 91 | ||
| AK391787.1 | - | 12 - 13 | 1578 | 91 | ||
| AK392919.1 | - | 7 - 8 | 791 | 91 | ||
| AK393422.1 | - | 12 - 13 | 1575 | 91 | ||
| AK398800.1 | - | 12 - 13 | 1619 | 91 | ||
| AK399390.1 | - | 6 - 7 | 629 | 91 | ||
Set Membership: |
|