Search
Search
| Gene Symbol: MYOF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073852.1 | XP_020929511.1 | 5819 | 62 | ||
| XM_021073853.1 | XP_020929512.1 | 5494 | 62 | |||
| XM_021073854.1 | XP_020929513.1 | 5780 | 62 | |||
| XM_021073855.1 | XP_020929514.1 | 5760 | 62 | |||
| XM_021073856.1 | XP_020929515.1 | 5722 | 62 | |||
| GenBank mRNA | AK236663.1 | - | 7 - 8 | 923 | 62 | |
| AK350841.1 | - | 7 - 8 | 899 | 62 | ||
Set Membership: |
|