Search
Search
| Gene Symbol: NCF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924811.4 | XP_001924846.2 | 842 | 74 | ||
| XM_013997542.2 | XP_013852996.1 | 86 | ||||
| XM_013997543.2 | XP_013852997.1 | 86 | ||||
| XM_021091535.1 | XP_020947194.1 | 2042 | 74 | |||
| GenBank mRNA | AK237150.1 | - | 7 - 8 | 816 | 74 | |
| AK237484.1 | - | 7 - 8 | 756 | 74 | ||
| AK393032.1 | - | 7 - 8 | 762 | 74 | ||
Set Membership: |
|