Search
Search

THUMPD2| Gene Symbol: THUMPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481230.4 | XP_003481278.2 | 1215 | 132 | ||
| XM_013987918.2 | XP_013843372.2 | 1456 | 132 | |||
| XM_013987919.2 | XP_013843373.2 | 1378 | 132 | |||
| GenBank mRNA | AK237372.1 | - | 9 - 10 | 1199 | 132 | |
| AK240141.1 | - | 9 - 10 | 1182 | 132 | ||
Set Membership: |
|