Search
Search
| Gene Symbol: NOP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005666511.3 | XP_005666568.2 | 2454 | 68 | ||
| GenBank mRNA | AK237695.1 | - | 8 - 9 | 964 | 68 | |
| AK350879.1 | - | 6 - 7 | 730 | 68 | ||
Set Membership: |
|