Search
Search
| Gene Symbol: CTPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013985938.2 | XP_013841392.1 | 1581 | 60 | ||
| XM_021080706.1 | XP_020936365.1 | 1800 | 60 | |||
| XM_021080707.1 | XP_020936366.1 | 1492 | 60 | |||
| XM_021080708.1 | XP_020936367.1 | 1399 | 60 | |||
| XM_021080709.1 | XP_020936368.1 | 1155 | 60 | |||
| GenBank mRNA | AK238015.1 | - | 10 - 11 | 1250 | 60 | |
| AK397843.1 | - | 10 - 11 | 1171 | 60 | ||
| AK400482.1 | - | 10 - 11 | 1365 | 60 | ||
Set Membership: |
|