Search
Search
| Gene Symbol: CHMP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354734.4 | XP_003354782.3 | 233 | 118 | ||
| GenBank mRNA | AK231203.1 | - | 1 - 2 | 116 | 118 | |
| AK232282.1 | - | 1 - 2 | 147 | 118 | ||
Set Membership: |
|