Search
Search
| Gene Symbol: BLNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001928233.5 | XP_001928268.1 | 391 | 62 | ||
| GenBank mRNA | AK231542.1 | - | 3 - 4 | 323 | 62 | |
| AK390973.1 | - | 3 - 4 | 333 | 62 | ||
Set Membership: |
|