Search
Search
| Gene Symbol: HACD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121729.4 | XP_003121777.2 | 882 | 96 | ||
| GenBank mRNA | AK238525.1 | - | 7 - 8 | 778 | 96 | |
| AK395723.1 | - | 7 - 8 | 819 | 96 | ||
Set Membership: |
|