Search
Search
| Gene Symbol: PLPP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924179.5 | XP_001924214.1 | 546 | 69 | ||
| GenBank mRNA | AK238542.1 | - | 5 - 6 | 550 | 69 | |
| AK238550.1 | - | 5 - 6 | 552 | 69 | ||
Set Membership: |
|