Search
Search
| Gene Symbol: CUL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131101.4 | XP_003131149.1 | 1832 | 71 | ||
| GenBank mRNA | AK238695.1 | - | 4 - 5 | 328 | 71 | |
| AK351500.1 | - | 3 - 4 | 300 | 71 | ||
Set Membership: |
|