Search
Search
| Gene Symbol: LXN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358644.4 | XP_003358692.1 | 770 | 100 | ||
| GenBank mRNA | AK238706.1 | - | 4 - 5 | 693 | 100 | |
| AK346816.1 | - | 4 - 5 | 692 | 100 | ||
| AK398073.1 | - | 4 - 5 | 711 | 100 | ||
Set Membership: |
|