Search
Search
| Gene Symbol: PARP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133932.5 | XP_003133980.2 | 517 | 67 | ||
| XM_021076746.1 | XP_020932405.1 | 442 | 67 | |||
| GenBank mRNA | AK238834.1 | - | 1 - 2 | 218 | 67 | |
Set Membership: |
|