Search
Search
| Gene Symbol: DARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133266.5 | XP_003133314.1 | 1282 | 73 | ||
| XM_021075843.1 | XP_020931502.1 | 1668 | 73 | |||
| GenBank mRNA | AK239116.1 | - | 11 - 12 | 1212 | 73 | |
| AK390493.1 | - | 11 - 12 | 1226 | 73 | ||
Set Membership: |
|