Search
Search
| Gene Symbol: DHX32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926580.6 | XP_001926615.2 | 2605 | 84 | ||
| XM_005671562.3 | XP_005671619.2 | 2623 | 84 | |||
| XM_021072595.1 | XP_020928254.1 | 2579 | 84 | |||
| XM_021072596.1 | XP_020928255.1 | 2946 | 84 | |||
| XM_021072598.1 | XP_020928257.1 | 2342 | 84 | |||
| XM_021072599.1 | XP_020928258.1 | 2467 | 84 | |||
| GenBank mRNA | AK239979.1 | - | 6 - 7 | 918 | 84 | |
| AK393530.1 | - | 7 - 8 | 1212 | 84 | ||
Set Membership: |
|