Search
Search
| Gene Symbol: VPS33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098980.1 | XP_020954639.1 | 843 | 78 | ||
| XM_021098981.1 | XP_020954640.1 | 458 | 78 | |||
| XM_021098982.1 | XP_020954641.1 | 458 | 78 | |||
| XM_021098983.1 | XP_020954642.1 | 828 | 78 | |||
| XM_021098984.1 | XP_020954643.1 | 594 | 78 | |||
| GenBank mRNA | AK343361.1 | - | 7 - 8 | 828 | 78 | |
Set Membership: |
|