Search
Search
| Gene Symbol: KLHL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021080422.1 | XP_020936081.1 | 676 | 76 | ||
| XM_021080423.1 | XP_020936082.1 | 717 | 76 | |||
| XM_021080424.1 | XP_020936083.1 | 583 | 76 | |||
| XM_021080425.1 | XP_020936084.1 | 314 | 76 | |||
| XM_021080426.1 | XP_020936085.1 | 234 | 76 | |||
| XM_021080427.1 | XP_020936086.1 | 411 | 76 | |||
| XM_021080428.1 | XP_020936087.1 | 472 | 76 | |||
| XM_021080429.1 | XP_020936088.1 | 676 | 76 | |||
| XM_021080430.1 | XP_020936089.1 | 479 | 76 | |||
| XM_021080431.1 | XP_020936090.1 | 590 | 76 | |||
| XM_021080432.1 | XP_020936091.1 | 613 | 76 | |||
| XM_021080433.1 | XP_020936092.1 | 558 | 76 | |||
| XM_021080434.1 | XP_020936093.1 | 437 | 76 | |||
| GenBank mRNA | AK343474.1 | - | 5 - 6 | 548 | 76 | |
| AK349310.1 | - | 3 - 4 | 495 | 76 | ||
Set Membership: |
|