Search
Search
| Gene Symbol: KLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122503.5 | XP_003122551.1 | 1829 | 89 | ||
| XM_005660664.3 | XP_005660721.1 | 1920 | 89 | |||
| XM_005660665.3 | XP_005660722.1 | 1881 | 89 | |||
| XM_021082807.1 | XP_020938466.1 | 2023 | 89 | |||
| XM_021082808.1 | XP_020938467.1 | 2042 | 89 | |||
| GenBank mRNA | AK343483.1 | - | 3 - 4 | 322 | 89 | |
| AK390086.1 | - | 13 - 14 | 1880 | 89 | ||
Set Membership: |
|