Search
Search
| Gene Symbol: BCORL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986421.2 | XP_013841875.1 | 7037 | 97 | ||
| XM_013986422.2 | XP_013841876.1 | 7120 | 97 | |||
| XM_013986424.2 | XP_013841878.1 | 6970 | 97 | |||
| XM_013986425.2 | XP_013841879.1 | 7625 | 97 | |||
| XM_013986426.2 | XP_013841880.1 | 7498 | 97 | |||
| XM_021080543.1 | XP_020936202.1 | 6992 | 97 | |||
| XM_021080545.1 | XP_020936204.1 | 7072 | 97 | |||
| XM_021080546.1 | XP_020936205.1 | 6833 | 97 | |||
| XM_021080547.1 | XP_020936206.1 | 6607 | 97 | |||
| GenBank mRNA | AK343574.1 | - | 1 | 909 | 97 | |
Set Membership: |
|