Search
Search
| Gene Symbol: FGF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135427.6 | XP_003135475.3 | 811 | 74 | ||
| XM_021080530.1 | XP_020936189.1 | 1240 | 74 | |||
| XM_021080531.1 | XP_020936190.1 | 1113 | 74 | |||
| XM_021080533.1 | XP_020936192.1 | 941 | 74 | |||
| XM_021080534.1 | XP_020936193.1 | 1071 | 74 | |||
| GenBank mRNA | AK343665.1 | - | 4 - 5 | 606 | 74 | |
Set Membership: |
|