Search
Search
| Gene Symbol: CEP152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659632.3 | XP_005659689.1 | 537 | 87 | ||
| XM_005659633.3 | XP_005659690.1 | 657 | 87 | |||
| XM_013993030.2 | XP_013848484.1 | 533 | 87 | |||
| XM_013993031.2 | XP_013848485.1 | 533 | 87 | |||
| XM_013993032.2 | XP_013848486.1 | 533 | 87 | |||
| XM_013993033.2 | XP_013848487.1 | 533 | 87 | |||
| XM_021095942.1 | XP_020951601.1 | 534 | 87 | |||
| XM_021095980.1 | XP_020951639.1 | 533 | 87 | |||
| GenBank mRNA | AK344402.1 | - | 4 - 5 | 376 | 87 | |
Set Membership: |
|