Search
Search

FASTKD2| Gene Symbol: FASTKD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005672124.3 | XP_005672181.1 | 2174 | 92 | ||
| XM_005672125.3 | XP_005672182.1 | 2145 | 92 | |||
| GenBank mRNA | AK344645.1 | - | 11 - 12 | 2110 | 92 | |
Set Membership: |
|